Would you like to log in again?
MUFFLER ASSEMBLY 6.4MM NPT 10U
Supplier: GAST MFG CORP
Muffler Assembly^ 10u. 6.4mm NPT. Replacement muffler assembly for Lubricated Rotary Vane Vacuum/Pressure Pumps (54907-059, -064; 54908-008, -012).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MUFFLER ASSEMBLY 6.4MM NPT 10U 54908-015 AB609B Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty