Would you like to log in again?
VACUUM PUMP UNMNTD BLTDRV 1STG
Supplier: WELCH VACUUM BY GARDNER DENVER
DUOSEAL* Unmounted Single-Stage Belt-Drive Vacuum Pump^ Unmounted pump without motor, base, or belt guard. Free air displacement: 35L/minute. Tubing: 11.1mm I.D. Oil capacity: 0.47L. Shipping weight: 23kg. Dimensions: 45Lx22.8Wx25.7Hcm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VACUUM PUMP UNMNTD BLTDRV 1STG 54972-706 1399 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty