Would you like to log in again?
VACUUM PUMP UNMNTD 2STG BLT-DR
Supplier: WELCH VACUUM BY GARDNER DENVER
DUOSEAL* Unmounted Two-Stage Belt-Drive Vacuum Pump^ Unmounted pump without motor, base, or belt guard. Elliptical bore design produces a high free air displacement, 500L/minute. Tubing: 41mm ID. Oil capacity: 1.18L.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VACUUM PUMP UNMNTD 2STG BLT-DR 54993-342 1397 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty