Would you like to log in again?
ELEMENT RPLCMNT CALCIUM HYDROX
Supplier: WELCH VACUUM BY GARDNER DENVER
Replacement Element^ An accessory for Acid Neutralization Trap (54994-092). Calcium hydroxide element. Element changes from white to bluish-transparent when spent.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ELEMENT RPLCMNT CALCIUM HYDROX 54994-074 1420E-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty