Would you like to log in again?
Acid Neutralization Trap, Welch®
Supplier: Gardner Denver
1420H-20
54994-092EA 653.78 USD InStock
54994-092
Acid Neutralization Trap, Welch®
Vacuum Traps
The Acid Neutralization Trap contains a large alkaline element cartridge to neutralize hydrous acids before they enter the vacuum pump
  • ISO NW flange connection
  • Calcium Hydroxide element
  • Configure with prior cold trap to reduce loading
The element changes from white to bluish-transparent when spent and can be observed through the transparent trap body.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty