Would you like to log in again?
VWR OIL VACUUM PUMP NO 19 1L
Supplier: VWR International
Superior, precision distilled oil based on durable and fine lubricating special crude oil stocks. Runs cooler in most mechanical pumps. Reduces thermal cracking for longer oil life. Good forepressures prolonged. Noticeable improvements in pumping cycle times. 55G.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR OIL VACUUM PUMP NO 19 1L 54996-061 Case of 12 (1/ Itm/Bx) Retrieving
Each(1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty