Would you like to log in again?
VWR® Vacuum Pump Oil No.19
Supplier: VWR
VWR®
418204 418200-55GL 418200-5GL 418203
54996-061CS 448.93 USD InStock
54996-061 54996-082 54996-128 54996-106
VWR® Vacuum Pump Oil No.19
Lubricants
This is a superior oil based upon special crude oil stocks noted for their durability and fine lubricating qualities.
  • Durable
  • Fine lubricating qualities
  • Clean oil runs coolly in mechanical pumps
  • Improves system lifetime
The oil is further enhanced through precision distilling, resulting in a cleaner oil that runs cooler in most mechanical pumps. Cooler operation reduces thermal cracking for longer oil life. Good forepressures are prolonged, reducing backstreaming and there are often noticeable improvements in pumping cycle times.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty