Would you like to log in again?
Precision® Vacuum Pump Oil, B+, Thermo Scientific
Supplier: Thermo Scientific
Precision®
3174013 3174014 3177864
54996-580EA 531.53 USD InStock
54996-580 54996-548 54996-503
Precision® Vacuum Pump Oil, B+, Thermo Scientific
Lubricants
Double vacuum distilled to remove light fractions
Low oil vapor pressure characteristics reduce backstreaming in vacuum systems and diffusion pumps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty