Would you like to log in again?
Female Pipe Adapters, Welch®, Gardner Denver
Supplier: Gardner Denver
506121 506142 506122
55009-149EA 123.81 USD InStock
55009-149 55009-176 55009-172
Female Pipe Adapters, Welch®, Gardner Denver
Fittings and Connectors
Female pipe adapters are made of stainless steel
It is compatible with Welch® thermocouple vacuum gauges 55008-186 and 55008-189 (see 55008-180 series) and other gauges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty