Would you like to log in again?
Rubber Hose Adapters, Welch®, Gardner Denver
Supplier: Gardner Denver
501283 501241 501251
55009-239EA 48.71 USD InStock
55009-239 55009-131 55009-152
Rubber Hose Adapters, Welch®, Gardner Denver
Fittings and Connectors
Converts ISO NW connection to a hose fitting
Made of aluminum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty