Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Photometry Reagents for 9000 Series Photometers, YSI
Supplier: YSI
YPM163 YPM166 YPM087 YPM168 YPM173 YPM268 YPM252 YPM177 YPM156 YPM179 YPM056 YPM258 YPM287 YPM162 YPM189 YPM188 YPM109 YPM155 YPM269 YPM284 YPM260 YPM148 YPM031 YPM175 YPM281 YPM186 YPM251 YPM011 YPM181 YPM254 YPM250 YPM060 YPM193 YPM130 YPM104 YPM105 YPM021 YPM152 YPM103 YPM266
55407-070EA 53 USD InStock
55407-070 55407-090 55407-150 55407-194 55407-094 55407-154 55407-110 55407-130 55407-098 55407-224 55407-104 55407-148 55407-168 55407-124 55407-228 55407-206 55407-128 55407-180 55407-062 55407-162 55407-082 55407-060 55407-220 55407-066 55407-088 55407-188 55407-086 55407-240 55407-164 55407-142 55407-120 55407-158 55407-114 55407-134 55407-212 55407-234 55407-118 55407-138 55407-216 55407-238
Photometry Reagents for 9000 Series Photometers, YSI
Microplate Readers
These photometry reagent starter kits for the 9000 Series Photometers include reagent tablets, instructions, and accessories for 50 tests
The replacement photometry reagents come in packs of 250 tests, except for fluoride (55407-108), nitrate (55407-152), phosphate LR (55407-170), silica (55407-184), and sulfide (55407-190), which come in packs of 200 tests.
The tablets have an indefinite shelf life and most require only a sample tube to perform test. No calibration from test to test or lot to lot is required.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty