Would you like to log in again?
Pocket Dosimeter, Fluke® Biomedical
Supplier: Fluke
06-912
55411-004EA 793.27 USD InStock
55411-004
Pocket Dosimeter, Fluke® Biomedical
Radiation Meters
Hermetically-sealed, direct-reading pocket dosimeter is a rugged, precision instrument designed for measuring accumulated quantities of gamma and x-radiation from 20keV to 2MeV
Meets ANSI specifications N13.5 and N322.
Can be clipped to a pocket or to the outside of a lead apron. To read, simply look through the eyepiece while pointing the unit toward any external light source. Exposure is determined by the position of a hairline fiber against a graduated scale. The dosimeter charger is used to re-zero. Electrical leakage is less than 0.5% of full scale for 24 hours at 50°C (122°F). Dimensions: 1.6dia.x11.4Lcm (5/8x41/2").
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty