Would you like to log in again?
CHARTS Z FOLD LEFT HAND PK10
Supplier: CARLTEX
Z-Fold Chart Paper^ For use with Data Acquisition Recorders. Length: 15m. Grid: 0 - 100 over 250mm. Zero left.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHARTS Z FOLD LEFT HAND PK10 55427-622 XR23 Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty