Would you like to log in again?
Max® and Max Lite® Earplugs, Honeywell Safety
Supplier: Honeywell Safety
Max® Max Lite®
LPF-30 MAX-1 MAX-30 MAX-1-D MAX-1-D LPF-30 MAX-30 LPF-1 MAX-1 LPF-1
55533-006CS 727.01 USD InStock
55533-006 CA55533-026 CA55533-004 55533-026 55533-004 CA55533-002 55533-002 CA55533-008 55533-008 CA55533-006
Max® and Max Lite® Earplugs, Honeywell Safety
Earplugs Disposable Earplugs
Pre-shaped foam earplugs feature a smooth outer skin for maximum user comfort.
  • Disposable, non-irritating and non-allergenic
  • Available with or without connecting cord
  • Max Lite® low-pressure self-adjusting foam fits smaller ear canals
Certifications: Meet ANSI S3.19 standard. CE marked. CSA certified.
Ordering information: Earplugs 55533-026 are bulk-packed for use in the Howard Leight dispenser (see 89013-668 series).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty