Would you like to log in again?
VWR® Heavy-Duty Single-Stage Gas Regulators
Supplier: VWR
VWR®
3001106 3001109 3001111 3001104 3001188 3001187 3001105 3001102 3001112 3001108 3001107 3001113 3001110 3001186 3001101
55850-270EA 676.7 USD InStock
55850-270 55850-260 55850-250 55850-240 55850-230 55850-275 55850-220 55850-277 55850-255 55850-200 55850-245 55850-235 55850-225 55850-215 55850-205
VWR® Heavy-Duty Single-Stage Gas Regulators
Gas Regulators
Regulators have a brass body and are suitable for a wide variety of applications where small changes in the gas delivery pressure will not degrade performance.
  • Brass body
  • Easy-to-read gauges
  • Large diaphragm ensures constant delivery pressure
  • Neoprene seat
  • Sintered metal inlet filter eliminates particulates
Gauges are easy to read
Note: For noncorrosive gases only. Use high-pressure hoses on regulators 55850-275, 55850-277, and 55850-270.
Large diaphragm ensures constant delivery pressure. Regulators with neoprene diaphragms have a valve, which closes with pressure for safety. Regulators with stainless steel diaphragms are not supplied with a valve. Particulates are eliminated from gas stream with sintered metal inlet filter.
Certifications: UL listed.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty