Would you like to log in again?
8000 Series Reusable Half Mask Respirators, Moldex®
Supplier: Moldex
8001 8003 8002
56219-044CS 539.95 USD InStock
56219-044 56219-042 56219-040
8000 Series Reusable Half Mask Respirators, Moldex®
Respirators Reusable Respirators Half Face Respirators
Half-mask features a soft, durable facepiece and an extra-wide sealing surface for a comfortable fit
Low profile allows a wide field of vision. Extra-large exhalation valve reduces hot air build-up and lowers breathing resistance. Strap adjustment is easy; loose ends are positioned out of line-of-sight. Low maintenance; no gaskets or yoke to replace. Snap-on cartridges for easy replacement.
Ordering information: Cartridges and filters must be ordered separately; please see accessory tab for additional information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty