Would you like to log in again?
L-Series Positive Pressure Headgear, 3M™
Supplier: 3M
7000126275 7000029620 7100077491 7000126275 7000126279 7000002080 7000126273 7000126279 7100077598 7100077491 7000029618 7000002080 7000029619 7000029618 7000029620 7100077598 7000002074 7000002074 7000126100 7000126273 7000029619 7000126100 7000042863
56221-406CS 1520.27 USD InStock
56221-406 CA56221-404 67100-608 56221-408 CA56221-406 56221-402 CA56221-400 CA56221-402 CA67100-608 56221-404 CA67100-694 67100-680 67100-630 67100-694 CA67100-680 CA67100-702 67100-644 CA67100-648 CA67100-630 CA56221-408 67100-648 67100-702 CA67100-644
L-Series Positive Pressure Headgear, 3M™
Respirators Respiratory Systems
Comfortable respirators are safe and cost-effective. No fit testing required. Positive pressure airflow has no breathing resistance and helps provide a higher level of respiratory protection. Wide-view lens provides better downward and peripheral vision. The lightweight headgear can be worn with facial hair, prescription eyewear, or safety glasses.
  • Comfortable, safe and cost effective; no fit testing required
  • Positive pressure airflow has no breathing resistance, provides higher level of respiratory protection
  • Wide-view lens provides better downward and peripheral vision
  • Can be worn with facial hair, prescription eyewear, or safety glasses
The L-500 series features a fully-adjustable, lightweight bumpcap
Supplied air is provided through either a high- or low-pressure assembly. The Vortex cooling assembly lets the worker regulate airflow and cool air by as much as 28°C (50°F). The Vortemp™ heating assembly lets the worker regulate airflow and warm air by as much as 28°C (50°F). The air regulating valve assembly allows the worker to regulate airflow to the headgear. The low-pressure assembly also regulates the airflow to the headgear.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Loose-fitting faceseal attaches to the face shield for respiratory protection up to 25x PEL (permissible exposure limit). Select from three styles: face shield, welding shield, or face shield/welding shield combination. Face shield protects face and eyes from debris, provides respiratory protection when down, and flips up when not needed. Welding shield/face shield combination features an inner shield to help reduce particles hitting the face and eyes during applications where the welding shield might be lifted. The inner shield provides respiratory protection when welding shield is up and allows workers to sight the work-piece before lowering the welding shield into place. The L-700 series features a lightweight hard hat that is fully adjustable and meets ANSI Class I plus Class II side-impact requirements for industrial protective headgear. A loose-fitting faceseal attaches to the face shield to provide respiratory protection up to 25x PEL. Select face shield, welding shield, or face shield/welding shield combination. The L-900 series features a helmet that meets ANSI Class I plus Class II side-impact requirements for industrial protective headgear. Model 53310-112 comes with a Tyvek® neck shroud, and model 53310-114 comes with a Nomex® neck shroud. Select face shield or face shield/welding shield combination. The L-900 series meets CAN/CSA Z94.1-92, Class G. Both powered and supplied air require breathing hose 67100-608. Cartridges are ordered separately.
Certifications: All systems are NIOSH approved when properly assembled.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty