Would you like to log in again?
Transpore™ Tape, 3M™
Supplier: 3M Healthcare
Transpore™
MMM1527-1 7000002796 7000002795 MMM1527-0
56222-104BX 48.37 USD InStock
56222-104 CA56222-106 CA11310-668 11310-668
Transpore™ Tape, 3M™
First Aid Supplies Wound Dressings
Transpore™ sergical tape is a latex-free, hypoallergenic, transparent and perforated plastic tape that offers strong adhesion.
  • Easy, straight, bi-directional tear allows clinicians to tailor tape to each use
  • Tape is also water-resistant
Transpore tape is easy to tear into very thin strips, and is easy to use even with gloves.
Ordering information: Tape is sold on a standard roll.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty