Would you like to log in again?
8576 P95 Particulate Respirator with Nuisance Level Acid Gas Relief, 3M™
Supplier: 3M
7000002061 7000002061
56222-364CS 928.08 USD InStock
CA56222-364 56222-364
8576 P95 Particulate Respirator with Nuisance Level Acid Gas Relief, 3M™
Respirators Disposable Respirators Particulate Respirators
Excellent choice for work settings that involve heat, humidity, and long wear periods.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Suggested uses include glass etching, chemical processing, paper processing, aluminum smelting, and brewing operations. Designed to provide comfortable, reliable worker protection against certain oil- and non-oil-based aerosol particles including those present with nuisance levels of acid gas such as sulphur dioxide, hydrogen fluoride, and chlorine. (Nuisance level acid gas refers to concentrations not exceeding the OSHA PEL or applicable government occupational exposure limits, whichever is lower.) Lightweight construction promotes greater worker comfort and contributes to increased wear time.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty