Would you like to log in again?
ADAPTER KIT BACK MNT'D HI-PRSR
Supplier: 3M OCCUPATIONAL HEALTH
Adapter Kit^ Includes 3M* Breathing Tube SA-2500 (56222-890), Waist Belt GVP-127(43300-210) and Air Regulator SA-1009(56222-880). This kit, with facepiece, filters/cartridges and air hose makes a 3M* Dual Airline Supplied Air Respirator.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER KIT BACK MNT'D HI-PRSR 56222-874 7000126161 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty