Would you like to log in again?
CONNECTOR ASMBLY F/2 AIR SYSTM
Supplier: 3M OCCUPATIONAL HEALTH
Connector Assembly^ Includes the 3M* Dual Airline Connector SA-1029 (56222-884) and the Waist Belt GVP-127 (43300-210). This assembly can be used with any 3M* Low Pressure Dual Airline Supplied Air Respirator.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONNECTOR ASMBLY F/2 AIR SYSTM 56222-882 7000126160 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty