Would you like to log in again?
Dual Airline Supplied Air Respirator Adapter Kits, 3M™
Supplier: 3M
7000126198 7000052039 7000126198 7000052039
56222-898CS 783.16 USD InStock
56222-898 CA56222-896 56222-896 CA56222-898
Dual Airline Supplied Air Respirator Adapter Kits, 3M™
Respirators Respiratory Systems
Dual airline breathing tubes deliver air to both sides of the facepiece for added comfort.
Combination dual airline provides both negative and positive pressure capability; negative pressure mode occurs automatically when air supply hose is disconnected.
Can be used with 3M™ 6000 series half-masks and full facepieces and 7800S series full facepieces. Half-masks fit under welding and other face shields. High-pressure regulator allows adjustment of airflow into the facepiece (also available in low pressure configuration).
Delivery information: Available with front- or back-mounted breathing tubes. Kit includes combination breathing tube, web belt and air regulator. Facepieces and cartridges/filters are ordered separately.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty