Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Air Purifying Dual Cartridge Respirators, Honeywell Safety
Supplier: Honeywell Safety
770030S 760008AS 54001S 770030M 54001 54001 770030M 760008AS 770030L 770030L 760008A 770030S 550030L 550030S 550030L 760008A 550030M 54001S 550030S 550030M
56223-224CS 344.55 USD InStock
56223-224 CA56223-222 CA56223-224 CA45000-754 45000-756 CA56222-948 45000-754 CA56222-946 56222-948 56223-220 CA45000-756 56223-222 CA56223-220 56222-952 CA56222-950 56222-950 CA56222-944 56222-946 CA56222-952 56222-944
Air Purifying Dual Cartridge Respirators, Honeywell Safety
Respirators Reusable Respirators Full Face Respirators
Comfortable half-mask or full facepiece respirators for use with North Safety Products cartridges and filters.
  • Reliable and latex-free respirators offer direct cartridge-to-facepiece connection
  • 7700 and 7600 series have nonallergenic silicone facepieces
  • 5500 and 5400 series have thermoplastic elastomer facepieces
  • NIOSH approved when used with North Safety Products cartridges
Interchangeable cartridges and filters provide air purifying protection for a variety of applications.
Half-mask respirators fit snugly while the cradle suspension system alleviates pressure points. Can be worn with safety glasses or goggles. Full facepiece respirators provide eye, face, and respiratory protection and are made of soft, flexible materials for optimum comfort.
The 7700 and 7600 series respirators have facepieces made of nonallergenic silicone; the 5500 and 5400 series respirators feature facepieces constructed of thermoplastic elastomer. Impact-resistant, wraparound polycarbonate lens permits 200° vision field. Full facepiece respirators include an oral/nasal cup. The 7600 series respirators include a speaking diaphragm.
NIOSH approved when used with North Safety Products cartridges and/or filters. Full facepiece lens meets ANSI Z87.1 standard for impact and penetration resistance. The eyeglass frame for the 7600 series respirators is constructed of sturdy metal and attaches to the lens of the facepiece with a suction cup.
Ordering information: For additional information on the accessory cartridges and filters, see 56222-957 series.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty