Would you like to log in again?
CARTRIDGE W/FLTR VPR/ACID PK2
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Organic Vapor/Acid Gas Cartridge with P100 Filter^ For organic vapor, chlorine, hydrogen chloride/fluoride, sulfur dioxide, chlorine dioxide, any particulates. 99.97% filter efficiency. Meets 42 CFR 84 requirements when used with North Safety air purifying dual cartridge respirators. NIOSH approved.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE W/FLTR VPR/ACID PK2 56222-980 7583P100L Case of 24 Retrieving
Pack of 2 Retrieving
Green Product
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty