Would you like to log in again?
Neoprene Glove Box Gloves, Honeywell Safety
Supplier: Honeywell Safety
8N1532AK/9Q 8N3032A/9Q 8N3032A/9Q 8NLL3032A/9Q 10N1532A/9Q 8N1532A/9Q 8NLL3032/10H 8N3032AK/10H 8N1532A/10H 7NLL3032/9Q 8NLL3032/9Q 10N3032A/9Q 8N3032AK/9Q 8N3032/8H 8N3032/8H 8N1532K/9Q 7N1532/9Q 5NLL3032A/9Q 8N1532/8H 5N3032/9Q 8N3032A/10H 8N1532/8H
56223-212PK 501.36 USD InStock
56223-212 CA56223-210 56223-147 56223-149 CA56223-147 56223-206 56223-208 32888-350 56223-150 56223-143 56223-210 CA56223-143 56223-145 82031-226 CA82031-314 82031-310 82031-312 82031-196 82031-262 82031-220 94002-630 82031-270
Neoprene Glove Box Gloves, Honeywell Safety
Gloves
Premium quality milled neoprene gloves offer high tensile strength and flexibility.
  • Seamless, one-piece construction provides maximum strength and integrity
  • Ideal for applications requiring good sensitivity and dexterity
  • Lead-loaded gloves contain 0.1 mm lead equivalent for attenuation of soft gamma radiation
Ordering information: Butyl, Hypalon®, Neoprene/Hypalon® blend, and Viton® glove box gloves are also available (see 82031-206 series).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty