Would you like to log in again?
Glove Box Gloves, Honeywell Safety
Supplier: Honeywell Safety
8B1532/8H 8B3032A/10H 8B3032A/9Q 8Y1532A/9Q 8B1532A/9Q 8Y3032A/9Q 8B1532/9Q 7NLY3032/9Q 7YLY3032A/9Q 8Y1532/8H 8B3032/9Q 8Y3032/10H 8Y3032/9Q 8Y1532A/9Q 7Y3032/9Q 8NY3032/10H 7Y1532A/9Q 8Y1532/9Q 8B1532A/10H 8NY3032/9Q 8Y1532A/10H 7YLY3032/9Q 8Y3032A/9Q 10B1532A/9Q 7NLY3032A/9Q 8B3032/9Q 8B1532/10H 8Y3032A/10H 5Y1532A/9Q 7B1532/9Q 8B3032/10H 8B1532/8H 8B1532A/9Q 7NY3032/9Q 8Y1532/9Q 8B3032/8H 7B1532A/9Q 8B3032A/9Q 8B1532/9Q 8B3032A/10H 5B3032A/9Q 8B3032/10H 8Y1532/8H 8Y3032A/9Q 5NY3032A/9Q 8Y3032/10H
56223-160PK 662.28 USD InStock
CA32940-073 56223-160 CA56223-192 32888-355 CA56223-152 CA63010-408 56223-188 CA56223-154 11000-570 11000-572 82031-288 CA82031-296 CA10746-666 CA82031-298 CA56223-190 CA82031-292 82031-286 CA11000-569 82031-282 32940-073 CA56223-202 CA56223-204 94002-638 56223-194 56223-152 32888-366 63010-408 56223-156 82031-236 82031-276 82031-254 82031-298 82031-232 56223-190 CA11000-573 82031-212 56223-192 11000-566 82031-272 82031-294 11000-568 82031-274 82031-252 82031-296 82031-290 82031-292
Glove Box Gloves, Honeywell Safety
Gloves
Butyl gloves provide the highest impermeability to water vapor, gases, and many toxic chemicals, including most missile fuels.
  • Wide variety of styles and sizes to meet diverse glove box applications
Hypalon® gloves offer outstanding abrasion and chemical resistance, including resistance to ozone and oxidizing chemicals. Hypalon® is naturally soft for greater comfort.
Lead-loaded gloves contain 0.1 mm lead equivalent for attenuation of soft gamma radiation.
Neoprene/Hypalon® blend gloves combine the flexibility and dexterity of neoprene with the ozone and chemical resistance of Hypalon®. Extra long life and positive protection under unusually severe conditions. Excellent resistance to ozone and oxidizing chemicals. White surface for good visibility; will not hide contamination.
Viton® gloves are designed for handling chlorinated and aromatic solvents. Provide superior resistance to PCBs. Excellent permeation resistance to many chlorinated, aliphatic, and aromatic hydrocarbons, and most oils and acids. Highly flame retardant. Excellent dexterity.
Flexible and sensitive, even at low temperatures. Resistant to oxygenated solvents and most oxidizing chemicals.
Ordering information: Neoprene glove box gloves are also available (see 82031-214 series).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty