Would you like to log in again?
Orbitron Rotators, Boekel Scientific
Supplier: Boekel Scientific
260250 260200
56261-508EA 1315.88 USD InStock
56261-508 35821-000
Orbitron Rotators, Boekel Scientific
Shakers and Mixers Rotating/Rocking Shakers
Gentle, uniform, low-foam mixing action suitable for immunoassays, hybridizations, mixing blood samples, coupling affinity chromatography media, and DNA extractions
Rotator I features a fixed speed of 20rpm and tilt of 23°. Rotator II features variable speed of 6–23rpm and tilt of 0–23°.
Compact unit has 30.5cm (12") square platform. Dimpled pad on platform accepts a variety of sizes of tubes simultaneously with no need for racks or clamps. Pad is removable for easy cleaning and test tube transport.
Ordering information: Rotator I is supplied with one dimpled pad. Rotator II is supplied with one dimpled pad and one flat, nonskid pad. Accessory tray kit includes a 20.3cm (8") platform, one dimpled pad, and one flat, nonskid pad.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty