Would you like to log in again?
Prinzing® Breaker Switch Lockouts, Brady®
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Prinzing®
45300 BL03-6 45305
56606-576PK 580.13 USD InStock
56606-576 CA56606-576 56606-572
Prinzing® Breaker Switch Lockouts, Brady®
Security Locks
Safely locks out breaker switches to prevent use during maintenance or service.
Includes notice not to lock breakers in the ON position. All lockouts accept #3 padlock.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty