Would you like to log in again?
VWR® Bottle Tote™ Safety Carriers
Supplier: Avantor
VWR® Bottle Tote™
B100-1 B-101 B-100 B-102 B102-1
56608-975EA 199.4 USD InStock
56608-975 56608-972 56608-970 56608-982 56608-980
VWR® Bottle Tote™ Safety Carriers
Carriers Bottle Carriers/Containers
Carriers are designed for the safe transport of acids, alkalies, and solvents.
  • High degree of tensile strength
  • Centering ring assures proper clearance for safe transport
  • Molded-in handle
High in tensile strength, carriers can be used in temperatures from -45 to 110°C (-49 to 230°F) and can be steam sterilized
The small carriers can accommodate 500 mL (16.9 oz.) or 1L (33.8 oz.) bottles. The large carriers can accommodate 2.5L (84.5 oz.) or 4L (135 oz.) bottles.
Centering ring in base assures a 2.54 cm (1") clearance between bottle and wall of carrier. Molded-in handle gives a comfortable grip.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty