Would you like to log in again?
Type I Nonmetallic Safety Cans, Justrite®
Supplier: Justrite
14561 14065 14160 14261
56609-592EA 179.8 USD InStock
56609-592 56609-621 56609-594 56609-595
Type I Nonmetallic Safety Cans, Justrite®
Safety Cans
High-density polyethylene cans are dent-, puncture-, and chip-resistant
  • Inert and resistant to tough acids, corrosives, and heat
  • Feature dual-density flame arrester, self-closing cap, and pressure relief mechanism
  • Current-carrying carbon insert imbedded in the can provides proper grounding
  • Insert prevents the creation of arc during filling or pouring
  • All cans are FM approved
  • Round cans are UL and C-UL listed
They feature a conductive current-carrying carbon insert embedded in the rib of the container, completing a ground path between the cover assembly and the flame arrester in the spout. When used with a grounding strip, this provides proper grounding and prevents the creation of an arc during filling or pouring. The self-closing cap, pressure relief mechanism, and dual-density flame arrester work together to prevent ignition sources without restricting liquid flow in or out of the can.
Cans are inert to resist tough acids and corrosives as well as heat.
Certifications: Cans are available with steel-plated or stainless steel hardware. FM approved. Round cans are UL and C-UL listed.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty