Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Plunger Cans, Justrite®
Supplier: Justrite
10308 10008 10208 10108 14018
56609-706EA 81.2 USD InStock
56609-706 56609-700 56609-702 56609-710 56609-708
Plunger Cans, Justrite®
Safety Cans
Plunger can helps moisten cleaning cloths or swabs quickly and easily
Upper pan fills when pushed down, and surplus liquid drains safely back into reservoir. Perforated pan screen serves as a flame arrester. Top rims have molded edge protectors for smooth, trouble-free moistening. Brass pump assembly. Available in terne plate or high-density polyethylene construction.
Certifications: FM approved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty