Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Bench Cans, Justrite®
Supplier: Justrite
10370 10575 10775 10175 10295 10375
56609-726EA 93.89 USD InStock
56609-726 56609-728 56609-722 56609-724 56609-730 56609-720
Bench Cans, Justrite®
Safety Cans
Terne plate steel bench can for convenient cleaning of small parts in solvents
Simply place part on spring-loaded, perforated dasher plate, press down and agitate, then release to return dasher to drain position. Plated steel dasher is an effective flame arrester. Hinged cover minimizes evaporation losses. Protected rim reduces possibility of hand injury. Item 56609-724 is a can with parts basket.
Certifications: FM approved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty