Would you like to log in again?
Nonmetallic Dispenser Cans, Justrite®
Supplier: Justrite
14004 14002 14010
56609-748EA 65.49 USD InStock
56609-748 56609-744 56609-740
Nonmetallic Dispenser Cans, Justrite®
Safety Cans
For easy one-hand dispensing of flammable liquids directly onto work
Dispensers 56609-744 and -748 are FM approved.
Brass dispenser valves are leakproof and self-closing to minimize spills and control excess. Rugged polyethylene body.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty