Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Safety Disposal Cans, Type 316 Stainless Steel, Eagle Manufacturing
Supplier: Eagle Mfg
1325 1323
56610-141EA 538.33 USD InStock
56610-141 56610-130
Safety Disposal Cans, Type 316 Stainless Steel, Eagle Manufacturing
Safety Cans
Designed for use as an intermediate station for liquid wastes.
  • Designed for use as an intermediate station for liquid wastes
The 10.1cm (4") I.D. opening permits these cans to be filled and emptied easily. Spout cap can be locked open for convenient emptying of many types of containers without use of a funnel. Each can is equipped with a flame arrester to reduce the possibility of ignition of contents by external spark. The cap assembly opens automatically to relieve excessive vapor pressure built up inside the can. The bottom of each can features a reinforced band of heavy-gauge steel for extra strength. Free-swinging handle maintains balance when can is carried, empty or full. With stainless steel fittings.
Certifications: FM approved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty