Would you like to log in again?
Oily Waste Cans, Polyethylene, Eagle Manufacturing
Supplier: Eagle Mfg
933FLY 935FL 937FLY 935FLY 933FL 937FL
56610-218EA 279.8 USD InStock
56610-218 56610-175 56610-220 56617-335 56617-330 56610-216
Oily Waste Cans, Polyethylene, Eagle Manufacturing
Bins Waste Bins
Cans are designed for the disposal of solvent and oily waste, rags, and other materials subject to spontaneous combustion.
  • Impact-resistant
  • Corrosion-resistant
  • Rustproof
Impervious to many liquid chemicals, solvents, and corrosives. A foot lever raises the lid which closes automatically after use.
FM approved.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty