Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Flammable Liquids Safety Storage Cabinets, Eagle Manufacturing
Supplier: Eagle MHC
1947X 1925X 1925X 1924X 1962X 4510X 1904X 1932X 6010X 1992X 3010X 1905X 1906X 1903X 9010X 1930X 1962XBEI 1945X
56610-390EA 1366.06 USD InStock
56610-390 56610-395 470202-550 56610-256 56610-245 56610-246 56610-301 56610-356 56610-254 56610-232 56610-200 56610-645 56610-643 56610-247 56610-258 56610-214 56610-248 56610-639
Flammable Liquids Safety Storage Cabinets, Eagle Manufacturing
Furniture Storage Cabinets
Storage cabinets constructed of 18-gauge steel with a caution yellow powder-coat finish with “FLAMMABLE–KEEP FIRE AWAY” in red lettering on the front.
  • Label is in three languages (English, Spanish, French)
  • The sides, top, bottom and doors are double-walled
  • 3.8 cm (1¹/₂") air space between walls
Cabinets with self-closing doors are equipped with a fusible link that automatically closes the door if the link melts due to fire.
Vents have 5 cm (2") pipe-thread fittings and are equipped with a fire baffle and cap. Cabinets are equipped with grounding attachments, three-point key lock on doors, 5 cm (2") raised, leakproof door sills, and four leveling feet. Each adjustable shelf is supported by four brackets.
Certifications: Conform to OSHA flammable liquid storage requirements and NFPA Code 30. Models with a capacity of 113.6 L (30 gal.) to 227.1 L (60 gal.) are FM approved.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty