Would you like to log in again?
Drum Cradle, Justrite®
Supplier: Justrite
08308 08800
56610-470EA 758.65 USD InStock
56610-470 56610-620
Drum Cradle, Justrite®
Carts
The drum siphon and self-closing faucet provide a simple solution for nearly complete drainage of liquids out of 113.6 L (30 gal.) and 208.2 L (55 gal.) drums, eliminating the need for dangerous tipping or rolling.
  • Cradle allows easy transport of drums weighing up to 363 kg (800 lbs.)
  • Loading bar grips drum chime to ensure smooth movement between vertical and horizontal positions
  • Designed to roll right into Justrite drum cabinets
  • Four wheels
Unique siphon action empties residual solvents from drums, typically down to the last few ounces. Siphon and faucet are constructed of solid cast brass.
Certifications: Cradle is FM approved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty