Would you like to log in again?
Opti-Klens™ I and Opti-Klens™ II Eyewashes, Desert Assembly
Supplier: Desert Assembly
Opti-Klens™
OK-IIREG TD-1 OK-I
56610-924CS 2799.02 USD InStock
56610-924 56610-935 56610-968
Opti-Klens™ I and Opti-Klens™ II Eyewashes, Desert Assembly
First Aid Supplies Eyewash Stations and Eyewash Bottles
Easy to install, no special plumbing required, these units will convert any faucet into an emergency eyewash station.
  • Opti-Klens™ II fills the need for a full face/eyewash
  • Delivers over 11.4 L/min. (3 gpm)
  • Push knob or turn faucet off to return to normal operation
Opti-Klens™ I has a threaded inlet that attaches to any standard faucet. Simply pull knob to activate. Water pressure holds eye wash in operation and leaves hands free.
The tubing diverter eliminates the need to remove an aerator from the faucet spout, attach a tubing barb, then reverse the procedure for regular use. Simply replace the aerator with the tubing diverter and leave it on. Pull button to fill containers as needed. Tubing diverter can be used with Opti-Klens™ on the same faucet.
Certifications: Meets ANSI standards for a face/eyewash fountain.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty