Would you like to log in again?
Fendall® Saline Eyewash Bottles, Honeywell Safety
Supplier: Honeywell Safety
32-000452-0000 32-000451-0000 32-000455-0000-H5
56611-118CS 203.85 USD InStock
56611-118 56611-124 56611-116
Fendall® Saline Eyewash Bottles, Honeywell Safety
First Aid Supplies Eyewash Stations and Eyewash Bottles
This bottle contains a sterile, buffered saline solution for emergency eye care.
  • Expiry dates and lot numbers for end-user protection
  • User-controlled volume delivery: one, four, and eight-ounce products
It delivers drops or an irrigating stream of solution, depending on the squeeze pressure applied to the bottle.
Each bottle features extended flow openings and twist-off tops for easy access in an emergency.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty