Would you like to log in again?
GLOVES POLYMER W/ NIT LG PK100
Supplier: CT INTERNATIONAL
Nitrile Co-Polymer Powder-Free Exam Gloves^ Large. Antistatic. 100% nitrile. Textured and tapered. Beaded cuff. Leak-tested. No latex proteins. White. Length: 30.5cm. Thickness: 5mil. Meets or exceeds ASTM D3578-95 Standards. Double-bagged. ISO Class 5 (FED-STD-209E Class 100/M3.5).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES POLYMER W/ NIT LG PK100 56613-862 NCP203 Case of 1000 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty