Would you like to log in again?
HyFlex® 70-400 Cut Resistant Gloves, Ansell
Supplier: Ansell
HyFlex®
242267 242086 242087 242266
56614-202CS 1404.42 USD InStock
56614-202 56614-200 47727-306 56614-204
HyFlex® 70-400 Cut Resistant Gloves, Ansell
Gloves
Type 70-400 cut-resistant gloves are knit from 100% Kevlar® continuous filament fiber.
  • Gloves are produced in a silicone-free environment
  • They have been prewashed to prevent shrinkage
These gloves are suitable for use in computer manufacturing, electronics and electrical assembly, and automotive paint room applications
They can also be used as cleanroom undergloves.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty