Would you like to log in again?
VAPOR MONITOR ORGANC BULK CS50
Supplier: 3M OCCUPATIONAL HEALTH
Vapor Monitor^ This monitor uses a single charcoal sorbent wafer for collecting organic vapors. Accuracy of +/-25% at a 95% confidence level for many workplace contaminants. Bulk packed. Analysis not included.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VAPOR MONITOR ORGANC BULK CS50 56614-744 7000126167 Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty