Would you like to log in again?
Maintenance Sorbents, 3M Company
Supplier: 3M
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7000051844 7000001942 7000126039 7000126020 7000126039 7000126019 7100134258 7000029594 7000126020 7000029595 7000001942 7000029594
56616-324CS 178.41 USD InStock
56616-324 CA56616-398 56616-322 CA56616-320 56616-397 56616-320 CA56616-395 56616-398 56616-395 67101-218 CA56616-391 56616-391
Maintenance Sorbents, 3M Company
Spill Control Materials
Maintenance sorbent mini-booms can be placed around the base of equipment to help contain fluids that may leak or spill.
  • For use on most non-aggressive liquids
  • Available in three lengths
  • Ideal for placing around equipment
  • Helps prevent leaks/spills from spreading to adjacent areas
  • Scrim on the outside of mini-boom identifies the use while containing particulate Sorbent material
Maintenance sorbent mini-booms can be placed around the base of equipment to help contain fluids that may leak or spill, preventing spread to adjacent work areas, One gallon capacity per four-foot Mini-Boom
Maintenance sorbent pillows are designed with a scrim that encases particulate sorbent material. Used in areas where greater capacity than a sorbent pad is desired. Small and lightweight, pillows are tear-resistant. This high capacity maintenance sorbent has a scrim on one side to increase strength and help eliminate linting. This conveniently sized pad allows the user to quickly spread sorbent on and in the path of a spill.
The maintenance sorbent rug is designed for use in industrial maintenance applications. It is designed to absorb liquids while enduring the punishment of being tread upon, sorbent rugs are both durable and versatile.
Four feet long by three inch diameter. Because of their tube-like shape, mini-booms are typically used to encircle spills and to contain leaks. A three-inch diameter scrim on the outside of the boom identifies the use while containing particulate sorbent material designed for the application.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty