Would you like to log in again?
Drum Tray, Eagle Manufacturing
Supplier: Eagle Mfg
1618 1614
56617-343EA 316.39 USD InStock
56617-343 56618-538
Drum Tray, Eagle Manufacturing
Drum Storage
Tray with reservoir keeps container away from spills
Ideal for use under drums during dispensing/filling operations. Holds up to 34L (9gal.) of spills when used with a 208.2L (55gal.) drum. Constructed of high-density polyethylene. Use drum tray dolly to easily transport drums between work areas without compromising spill protection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty