Would you like to log in again?
Biohazard Waste Containers, Justrite®
Supplier: Justrite
05910 05930
56617-832EA 389.08 USD InStock
56617-832 56617-830
Biohazard Waste Containers, Justrite®
Bins Waste Bins
For temporary collection of hazardous biological waste
  • For segregation of infectious waste
  • Leakproof, sturdy metal construction
  • Simple, foot-operated, self-closing lid to reduce contamination
  • Biohazard bag is held securely around the circumference of the can
Caution: Not to be used with sharps.
May be used for temporary storage of contaminated laundry as well as other regulated waste.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty