Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Miller DuraFlex® Ultra and Python® Ultra Harnesses, Honeywell Safety
Supplier: Miller by Honeywell
DuraFlex® Python™
E650-4/UGN E650/UGN E650-58/UGN P950QC/UGN P950QC-7/UGN
56619-022EA 632.06 USD InStock
56619-022 56619-012 56619-014 56619-018 56619-020
Miller DuraFlex® Ultra and Python® Ultra Harnesses, Honeywell Safety
Fall Protection Fall Harnesses
Harnesses feature sleek, quick-connect chest and leg strap buckles for easy donning
  • Sleek, quick-connect chest and leg strap buckles for easy donning
  • Buckles have a dual-tab release mechanism to prevent accidental opening
  • Comfort-Touch back D-ring pad is designed with woven, breathable material for cooler extended-wear comfort
  • Pad orients harness for quick, tangle-free donning
The DuraFlex® Ultra harnesses feature stretchable webbing that provides greater comfort and improves safety. The DuraFlex Python® Ultra harnesses feature cushioned tubular webbing in the shoulder straps to minimize discomfort around the neck and shoulder when user is required to wear a tool belt. Universal size fits large and X-large.
Buckles have a dual-tab release mechanism to prevent accidental opening. The Comfort-Touch back D-ring pad is designed with woven, breathable material for cooler extended wear comfort. Pad orients harness for quick, tangle-free donning.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty