Would you like to log in again?
POWERCOAT® Disposable Tri-polymer Gloves, Honeywell Safety
Supplier: Honeywell Safety
PowerCoat®
PSD-TRIP-XL PSD-TRIP-M PSD-TRIP-L PSD-TRIP-M PSD-TRIP-S PSD-TRIP-S PSD-TRIP-L
56619-980BX 67.43 USD InStock
56619-980 CA56619-980 CA56619-982 56619-976 56619-978 CA56619-976 CA56619-978
POWERCOAT® Disposable Tri-polymer Gloves, Honeywell Safety
Gloves
These are tri-polymer disposable gloves with a blend of latex, neoprene and nitrile that provides excellent chemical and liquid protection.
  • Powder-free for reduced product contamination
  • Finger tips have a rough finish for added grip in wet applications
Ideal for use in agriculture, chemical, oil and gas industries.
Certifications: According to ANSI.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty