Would you like to log in again?
PIG® Sump Skimmer, New Pig
Supplier: New Pig
SKM403
56620-022CS 218.97 USD InStock
56620-022
PIG® Sump Skimmer, New Pig
Spill Control Materials
Simply lower this floating skimmer into any water-based liquid to absorb oils without taking in water.
  • Absorbs and retains oils and oil-based liquids, including lubricants and fuels, without taking in a drop of water
  • Bright white color makes absorbed oil easier to see; clearly shows saturation level
  • Can be incinerated after use to reduce waste or for fuels blending
  • Spaghetti strip filler is designed for capturing thick oils or other viscous liquids
  • Tough outer mesh is UV resistant for long-term outdoor use without degradation; lets fluids easily pass through to filler material
Excellent for soaking up unwanted oil-based fluids that collect in sumps, tanks and bilges. Polypropylene loop can be attached to rope (not included) for easy deployment and retrieval.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty