Would you like to log in again?
PIG® HazMat Chemical Absorbent Pillow in Dispenser Box, New Pig
Supplier: New Pig
PIL306 PIL302
56620-028CS 245.87 USD InStock
56620-028 56620-029
PIG® HazMat Chemical Absorbent Pillow in Dispenser Box, New Pig
Spill Control Materials
  • Quickly absorb highly concentrated corrosive liquids, including 98% sulfuric acid and 30% sodium hydroxide
  • Highly absorbent polypropylene will not degrade upon contact with harsh or aggressive liquids
  • Made of 100% chemical-resistant skin and fast-wicking filler
  • Large surface area and high capacity to quickly soak up liquids
Absorbs the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide. Chemical-resistant pillow won't degrade or cause a dangerous reaction upon contact with corrosive spills. Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up liquids; great for adding absorbency during spill response. Polypropylene skin resists chemicals and tearing; reduces dust and holds in liquid, even when saturated. Polypropylene filler is highly absorbent for containing corrosive or reactive spills.
Absorbs and retains most acids, bases and unknown liquids. Pink color is easily distinguishable to assure workers use the correct pillow during a spill emergency. Can be incinerated after use to reduce waste. Dispenser box makes pillows easy to access, carry and store while protecting contents from dirt and moisture.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty