Would you like to log in again?
PIG® Oil-Only Absorbent Mat Roll, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG®
MAT425 MAT401 MAT419 MAT462 MAT424 MAT441 MAT461
56620-066EA 183.87 USD InStock
56620-066 89073-844 89205-906 89073-856 89073-850 89073-858 89073-848
PIG® Oil-Only Absorbent Mat Roll, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to give PIG Mat excellent strength; won't rip, tear or fray even when saturated
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job
  • Rolls are ideal for covering large areas and soaking up leaks and overspray
  • Easy-tear perforations let you take only what you need, so you use less mat and save money
Can be wrung out and incinerated after use to reduce waste or for fuels blending. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Rolls are ideal for covering large areas and soaking up leaks and overspray. Easy-tear perforations let you take only what you need, so you use less mat and save money.
Heavyweight construction is highly durable and absorbent for demanding tasks and large-volume cleanups. Mediumweight construction is versatile for general, everyday tasks and cleanups. Light-weight construction is economical for small tasks and cleanups.
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave behind liquids or fiber residue. Absorbs and retains oils and oil-based liquids - including lubricants and fuels - without taking in a drop of water. Bright white color makes absorbed oil easier to see; draws attention to machine leaks and clearly shows saturation level during spill response. Floats to clean up spills on water
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty